Electrode stand with swivel arm, Eutech
Leverandør: EUTECH
ECPHELSTDC
EUTEECPHELSTDCEA 1500 NOK
EUTEECPHELSTDC
Electrode stand with swivel arm, Eutech
Elektrodestativ
This electrode stand with swivel arm allows convenient hands-free operation. Holds up to four standard size electrodes and one ATC probe.
  • Weighted 6-inch diameter base allows users to pivot holder 360° for more flexibility
  • Arm allows users to move electrode side to side or up and down while keeping electrode at a constant vertical angle