Anti-PENK Goat Polyclonal Antibody

Leverandør: Everest Biotech
EB08196
EVBIEB08196EA 3975 NOK
EVBIEB08196
Anti-PENK Goat Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: PENK (proenkephalin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: internal
Host: Goat
Isotype: IgG
Reactivity: Mouse, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR