Hand cleansing creams, Kresto® paint liquid
Leverandør: Deb-STOKO
22306
EVON22306EA 3200 NOK
EVON22306
Hand cleansing creams, Kresto® paint liquid
Hudpleie Såper og hud rensemidler
The special hand cleanser with very powerful cleansing action contains mild solvents. It removes very stubborn and extremely heavy-duty contamination like lacquers, resins, adhesives, oil-based paints.
  • Scrubbing agent-free
  • As the formulation is concentrated, only a small amount is required to remove contaminants
  • Solvent-free, colourant-free, preservative-free, perfumed
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904