Foam hand sanitiser dispenser, InstantFOAM TouchFREE
Leverandør: Deb-STOKO
IFSTF2MD-UK
EVONIFSTF2MD-UKEA 1225 NOK
EVONIFSTF2MD-UK
Foam hand sanitiser dispenser, InstantFOAM TouchFREE
Såpedispensere og Lotion Såpedispensere
Energy-efficient touch-free dispensing system for use with foam hand cleansers.
  • Minimises any risk of cross-contamination
  • 4 D-cell batteries provide up to five years of operation
  • Infrared sensing technology to prevent misfiring
  • BioCote® protection antimicrobial agent inhibits the growth of bacteria and moulds
  • Translucent covers provide visibility of the cartridge without opening the dispenser
  • Dispenser can be locked and opened with the special key provided, can also be easily converted to a keyless, non locking operation
  • For wall-mounting
Suitable forMore Product Information