Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 5 µm
Leverandør: Avantor
EXL-1214-2502UEA 0 NOK
EXL-1214-2502U EXL-1214-2546U ACE12140103GD EXL-1214-0502U EXL-1214-0546U EXL-1214-1003U EXL-1214-1246U EXL-1214-1502U EXL-1214-1546U
Avantor® ACE® Excel® NH₂, HPLC/UHPLC Columns, Analytical, 5 µm
Kromatografikolonner
These stainless steel columns with proprietary bonding technology provide high robustness with excellent column lifetimes.
  • Usable in reversed phase, normal phase, weak anion-exchange and HILIC modes
  • Ideal for the analysis of sugars
  • Ultra-inert, high efficiency 1,7 µm, 3 µm and 5 µm particles for optimum peak shape
  • Method validation kits contain three different batches of the same phase - available on request