Deep well collection plates, 96-Well, 2,2 ml
Leverandør: OMEGA BIO-TEK
EZ9602-02
EZ9602-02EA 2450 NOK
EZ9602-02
Deep well collection plates, 96-Well, 2,2 ml
Mikroplater HTS-mikroplater
96-well PP collection plates with 2,0 ml well volume for lysis, sample cultivation, and flow-through collection. This square well plate allows for larger volume than the equivalent round well plate in a standard microplate format.
  • Collection plate fits well with most robotic work stations
  • Round bottom microplate fits magnetic separation device (MSD-1)
  • Manufactured from high quality polypropylene (PP) for excellent temperature and chemical resistance