FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) fluorescent labeling reagent

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: 5-isothiocyanatofluorescein, Fluorescein 5(6)-isothiocyanate, Fluorescein Isothiocyanate, Isomer I

MQ200

F1628-1G F1628-250MG
F1628-1GEA 1670 NOK
F1628-1G F1628-250MG
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I) fluorescent labeling reagent
FITC (fluorescein-5-isothiocyanate, fluorescein isothiocyanate isomer I)

Fluorescein isothiocyanate isomer I–Celite® has been conjugated with antibody for fluorescence imaging in cells and flow cytometry analysis.

Formel: C₂₁H₁₁NO₅S
MW: 389.381 g/mol
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00005063
CAS-nummer: 3326-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f6d7eb4a5209f23db32f7515ee463d14d1331fde&vid=0b9d42251b68d49ad627398f19043d0c57705b0e&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Yellow to Orange
Appearance (Form) Powder
Assay
FTIC Content
8.0 - 12.0 %
Suitability
Suitable for use in Protein Labeling
Suitable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR