O-Dianisidine dihydrochloride suitable for use in glucose determination, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: 3,3'-Dimethoxybenzidine dihydrochloride

M-Clarity™ quality level = MQ200

F5803-50MG
F5803-50MGEA 932 NOK
F5803-50MG
O-Dianisidine dihydrochloride suitable for use in glucose determination, Sigma-Aldrich®
O-Dianisidine dihydrochloride

A preweighed vial containing 50 mg o-Dianisidine. o-Dianisidine (3,3′-dimethoxybenzidine) is a peroxidase substrate suitable for use in ELISA procedures. This substrate produces a soluble end product that is yellow-orange in color and can be read spectrophotometrically at 405 nm. The reaction may be stopped with 5 M HCl.


  • Suitable for use in the glucose determination


It is suitable reagent used in the following studies:
- As substrate for the determination of serum ceruloplasmin from its oxidase activity.
- Determination of total antioxidant response (TAR) against potent free radical reactions by a novel, colorimetric and fully automated method.
- As colorimetric indicator for the simultaneous semi-quantitative estimation of glucose, lactate and uric acid on paper-based microfluidic devices (microPAD).
- Myeloperoxidase activity assay.


NB: Probably carcinogenic.

Formel: C₁₄H₁₈Cl₂N₂O₂
MW: 317.21
Smeltepunkt: 268 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00012488
CAS-nummer: 20325-40-0
EINECS: 243-737-5
FN: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d4a9b8329605d5166121fede8c21b49f99bcba8b&vid=5d66ccc365ce0cc870fc232369afdb4f15a71682&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) White
Appearance (Form) Crystals
Solubility (Turbidity), 50 mg vial reconstituted with 20 ml of purified water Clear
Solubility (Colour), 50 mg vial reconstituted with 20 ml of purified water Colourless
Solubility (Form), 50 mg vial reconstituted with 20 ml of purified water Solution
pH 6.95 - 7.05
Glucose Recovery, Recovery of 3 mg/ml NIST Glucose Standard 2.80 - 3.50 mg/ml
Absorbance A450 Blank ≤0.05
Completion Time ≤30 min


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR