Fast Blue RR Salt, capsules, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: Azoic Diazo No. 24, 4-Benzoylamino-2,5-dimethoxybenzenediazonium chloride hemi(zinc chloride) salt

MQ500

FBS25-10CAP
FBS25-10CAPEA 1740 NOK
FBS25-10CAP
Fast Blue RR Salt, capsules, Sigma-Aldrich®
Fast Blue RR Salt

Fast Blue RR is part of the azo coupling system and is required for the staining of hydrolytic enzymes. In the presence of naphthol, it results in the formation of an insoluble coloured precipitate at the site of enzyme activity.

Formel: C₁₅H₁₄ClN₃O₃
MW: 387.89 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00074762
CAS-nummer: 14726-29-5
EINECS: 238-777-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bdf65bd9fff60d7744c05de55bfab9d223c6e389&vid=bdf65bd9fff60d7744c05de55bfab9d223c6e389&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Colour) Yellow and Yellow-Green
Appearance (Form) Powder
Performance, When staining with Sigma-Aldrich Procedure No. 85, cells stain in accordance with the package insert. Conforms


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR