Restriction enzymes, SalI, FastDigest™, Fermentas
Leverandør: Thermo Fisher Scientific
FD0644
FD0644EA 770 NOK
FD0644
Restriction enzymes, SalI, FastDigest™, Fermentas
Nukleinsyre reagenser Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications
Designed to complete digestion in 5 to 15 minutes.