Restriction enzymes, XhoI, FastDigest™, Fermentas
Leverandør: Thermo Fisher Scientific
FD0694 FD0695
FD0694EA 670 NOK
FD0694 FD0695
Restriction enzymes, XhoI, FastDigest™, Fermentas
Nukleinsyre reagenser Restriction Enzymes
FastDigest™ XhoI is one of an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest™ and FastDigest™ Green reaction buffers.
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5 to 15 minutes
  • Direct loading on gels
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon
Enzymnavn XhoI
Kilde
Overhang type 5' overhang
Kompatibel buffer FastDigest Green Buffer or FastDigest Buffer (colorless)