Restriction enzymes, BpiI, FastDigest™, fermentas
Leverandør: Thermo Fisher Scientific
FD1014
FD1014EA 490 NOK
FD1014
Restriction enzymes, BpiI, FastDigest™, fermentas
Nukleinsyre reagenser Restriction Enzymes
Thermo Scientific™ FastDigest™ enzymes are an advanced line of restriction enzymes for rapid DNA digestion.
  • 100% activity of all FastDigest™ enzymes in one universal reaction buffer
  • Designed to complete digestion in 5 to 15 minutes
  • Direct loading on gels with FastDigest™ Green buffer
  • 100% buffer compatibility with downstream applications