Restriction enzymes, FastDigest SdaI, Fermentas

Leverandør: Thermo Fisher Scientific

FD1194
FD1194EA 740 NOK
FD1194
Restriction enzymes, FastDigest SdaI, Fermentas
Nukleinsyre reagenser Restriction Enzymes

FastDigest SdaI is an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest green reaction buffers. This enzyme recognises CCTGCA^GG site and cuts best at 37 °C in 5 to 15 minutes using universal FastDigest buffer.


  • Direct loading on gels and 176 FastDigest enzymes available
  • No star activity
  • Buffer compatibility of 100% with downstream applications
  • Methylation sensitivity: not CpG methylation-sensitive, dam methylation-sensitive and dcm methylation-sensitive


The universal buffer allows rapid single-, double-, or multiple DNA digestion within 5 to 15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps.


Applications include molecular cloning, restriction site mapping, genotyping, southern blot, restriction fragment length polymorphism (RFLP) and SNP analysis.


NB: For research use only. Not for use in diagnostic procedures.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR