KingFisher™ Presto Purification System
Leverandør: Thermo Fisher Scientific
5400830
FINN5400830EA 0 NOK
FINN5400830
KingFisher™ Presto Purification System
Renseaparat for nuklein syrer
Use the Thermo Scientific KingFisher Presto as part of an automated purification system with a liquid handler for plate filling in high-throughput laboratories.
  • Volume range of 50 to 5000 μl
  • Minimised size for easy fitting to automation systems
  • Heating capability for application requiring elevated temperatures
  • Easy connection to several liquid handling instruments
  • Flexible system for wide variety of applications
Utilising proven KingFisher magnetic particle-based technology, the KingFisher Presto provides high-quality yields of target nucleic acidsMore Product Information