Formalin 10%, pink, Q Path®, buffered

Leverandør: VWR Q-Path Chemicals
Danger

FOR0023AF59001EA 945 NOK
FOR0023AF59001 FOR0153AF59001 FOR0063AF59001
Formalin 10%, pink, Q Path®, buffered
Formalin

Formalin pink ready to use fixative for tissue samples. Pots can be used for storage and transport.


Formaldehyde 4% is approximately equal to Formalin 10%

Formel: CH₂O
Tetthet: 1,013 g/cm³ (20 °C)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00003274
CAS-nummer: 50-00-0
EINECS: 200-001-8
IVDLogo

Bestill nå

Spesifikasjon Testresultater

Appearance Pink liquid
Test formaldehyde Passes test
Density (20/4) 1,012 - 1,014
pH (20°C) 6,8 - 7,2

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR