Glucose assay reagent, Supelco®
Leverandør: Merck
G3293-50ML G3293-20ML
G3293-50MLEA 1575 NOK
G3293-50ML G3293-20ML
Glucose assay reagent, Supelco®
Mat og drikkevare testsystemer
Activity <0,05% resulting from cross-reactivity with the following substrates: fructose, dextran, sedoheptulose, arabinose, galactose, xylose, trehalose, mannose, sorbitol, fucose or ribose.
For the quantitative determination of glucose in food and other materials utilizing the hexokinase-glucose 6-phosphate dehydrogenase enzymatic assay. Reconstitute with deionised water using volume indicated by package size (20 or 50 ml).
Forpakning: 20, 50 ml in serum bottle.
Bestill nå
SPESIFIKASJONER
Beskrivelse Glucose Assay Reagent