Giemsa's stain, Technical Grade used as a blood stain, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Warning

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

M-Clarity™ quality level = MQ100

G4507-25G G4507-5G
G4507-25GEA 4690 NOK
G4507-25G G4507-5G
Giemsa's stain, Technical Grade used as a blood stain, Sigma-Aldrich®
Giemsa's stain

Giemsa stain is a versatile differential stain that is widely used in hematology for blood and bone marrow specimens, clinical cytology specimens, bacteriology, histological biopsies, and tumor samples. Giemsa's staining solution is composed of methylene blue, azure, and eosin. It is a major histological stain due to its unique staining of chromatin, nuclear membranes, and cytoplasmic elements. The stain is frequently used in combination with other dye solutions: May-Grünwald’s solution for Pappenheim (MGG) and Wright-Giemsa.


  • Giemsa stain has been used for the staining of cells and blood films

Smeltepunkt: 300 °C (lit.)
Lagringstemperatur: Room Temperature
MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
EINECS: 257-438-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=65a3dde4b74ec23e6f527c0ce338d61d470c8d26&vid=c7baef9be683f2300682a16b9432418e0a4311a0&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Appearance (Color) Dark to Very Dark Green and Black
Appearance (Form) Powder
Solubility (Color) 4 mg plus 4 ml of Ethanol
Wavelength (1) (UV) 645 - 655 nm
Solvent (UV) Methanol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR