Permanent cryogenic labels for laser printers, US letter size
Leverandør: GA
CL-15T1-OR CL-15T1-PI
GAINCL-15T1-OREA 830 NOK
GAINCL-15T1-OR GAINCL-15T1-PI
Permanent cryogenic labels for laser printers, US letter size
Etiketter
Cryo-LazrTAG™ laser printable cryogenic labels are designed for the permanent identification of cryo vials and tubes, microtubes, blood collection tubes, cryo boxes, freezer boxes, cell culture plates, microplates, and other cryogenic and freezer containers.
  • Strong permanent adhesive ensures labels will not wrinkle or peel
  • Printable with all common desktop laser printer brands (US Letter Size: 21,5×27,9 cm)
  • Withstand a wide temperature range (–196 to 120 °C)
  • Available in a wide assortment of colors, sizes, and configurations
These labels are designedMore Product Information