Jolting volumeter
Leverandør: Gerber Instruments AG
09.0220
GEAG09.0220EA 0 NOK
GEAG09.0220
Jolting volumeter
Densitetsmålere
Volumeter designed for determination of specific weight of a jolted sample.
Automatic apparatus with up to 100 strokes in 30 seconds.