Human Serum Albumin (HSA) Solution (25%), 50 ml

Leverandør: GEMINI BIO-PRODUCTS
800-120
GEMB800-120EA 2875 NOK
GEMB800-120
Human Serum Albumin (HSA) Solution (25%), 50 ml
Serum
This diagnostic-grade product is an aqueous solution containing 25% human albumin (w/v).

It is prepared by cold-alcohol fractionation from pooled human plasma obtained from venous blood. The USP Pharmacopeia requires that all fractionators add sodium caprylate and sodium tryptophanate as a preservative.

Leveringsinformasjon: Ships on dry ice. Store frozen at −20 to −10 °C.

NB: For cell culture, further manufacturing and research use only. Not for direct therapeutic use.
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR