Protein A and G Antibody Purification Kits
Leverandør: G Bioscienes
786-283 786-284 786-828 786-827 786-826 786-825 786-833 786-831 786-830 786-829 786-824 786-834 786-832 786-835
786-283EA 11950 NOK
786-283 786-284 GENO786-828 GENO786-827 GENO786-826 GENO786-825 GENO786-833 GENO786-831 GENO786-830 GENO786-829 GENO786-824 GENO786-834 GENO786-832 GENO786-835
Protein A and G Antibody Purification Kits
Proteinrense Protein Purification Kits
Immobilised Protein A and G are designed for binding the constant domains of immunoglobulin (Ig) molecules. Immobilised Protein A is coupled to 6% highly cross-linked agarose beads by a proprietary coupling method that provides high coupling efficiency for immunoglobulins and minimal protein A leaching. The binding capacity of Protein A is 18 to 43 mg Human IgG per ml resin.
Immobilised Protein G is a modified form of Streptococcal group G so that it does not bind to albumin. Protein G is coupled to 4% cross-linked agarose beads by a proprietary coupling method that provides high coupling efficiency for Ig and minimal protein G leaching. The binding capacity of Protein G is >20 mg Human IgG per ml resin.
Bestillingsinformasjon: Immobilised Protein A and G Resin are available as resin-alone, prepacked columns or supplied in kit formats containing columns, wash and elution buffers.