Anti-Trx-tag Mouse Monoclonal Antibody

Leverandør: Genscript
A00180-40
GENSA00180-40EA 2025 NOK
GENSA00180-40
Anti-Trx-tag Mouse Monoclonal Antibody
Antistoffer
Type: Tag
Antigen: Trx-tag
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a (Mouse)
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR