Anti-GST Mouse Monoclonal Antibody (Biotin)

Leverandør: Genscript
A00867-200
GENSA00867-200EA 6725 NOK
GENSA00867-200
Anti-GST Mouse Monoclonal Antibody (Biotin)
Antistoffer
Type: Tag
Antigen: GST
Clonality: monoclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR