Anti-PEG Mouse Monoclonal Antibody

Leverandør: Genscript
A01795-100
GENSA01795-100EA 3200 NOK
GENSA01795-100
Anti-PEG Mouse Monoclonal Antibody
Antistoffer
Type: Tag
Antigen: PEG
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol PEG
Environmentally Preferable
Antistofftype Tag
Klonalitet Monoclonal
Konjugering Unconjugated
Reaktivitet
Vert Mouse
Isotype
Western blot Yes
ELISA Yes
ImmunoKjemi Yes
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR