Anti-DM1 Mouse Monoclonal Antibody [clone: 15L2]
Leverandør: Genscript
A02223-40
GENSA02223-40EA 3450 NOK
GENSA02223-40
Anti-DM1 Mouse Monoclonal Antibody [clone: 15L2]
Antistoffer
The DM1, also known as mertansine, is a microtubule polymerisation inhibitor. Trastuzumab emtansine (T-DM1), composed of trastuzumab conjugated to DM1, is a type of targeted cancer drug for breast cancer treatment. T-DM1 is targeted toward HER2 (ERBB2) positive breast cancer.
  • High specificity
  • High affinity
  • High stability