Taq DNA polymerase, 1000 U
Leverandør: Genscript
E00008
GENSE00008EA 1300 NOK
GENSE00008
Taq DNA polymerase, 1000 U
Nukleinsyre reagenser End-point PCR Enzymes and Kits
Taq DNA polymerase is a thermostable DNA polymerase isolated from an E. coli strain that carries the Taq DNA polymerase gene.
  • Used for PCR reactions
  • Terminal transferase activity
  • High purity
One unit is the amount of enzyme that can incorporate 10 nmol of dNTP into acid-insoluble material in 30 minutes at 74 °C.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon