Affinity purification resins, anti-His, GenScript®

Leverandør: Genscript

L00439-1 L00439-5
GENSL00439-1EA 8025 NOK
GENSL00439-1 GENSL00439-5
Affinity purification resins, anti-His, GenScript®
Proteinrense Protein Purification Resins

Anti-His affinity resins are designed for purification of His-tagged protein. Composed of agarose and conjugated THE™ anti-His monoclonal antibody, they are compatible with most commonly used reagents at certain concentrations.


  • Beads structure: 4% agarose
  • Beads size: 45 to 165 μm, average 90 μm
  • Binding capacity: greater than 3 mg of human C-reactive protein per ml of settled resin
  • Specificity: N-terminal/C-terminal/internal/4 x His/5 x His/6 x His-tagged protein
  • Storage: 2 to 8 °C (avoid freezing the resin)
  • Binding condition: Tris or borate-buffered saline (pH 8 to 8,5) with 1 to 2 mM calcium chloride
  • Elution methods: Acid, alkaline and peptide competitive elutions


His-tagged protein in the crude samples can bind to the anti-His monoclonal antibodies coupled with the agarose, and the non specific proteins can be eliminated by washing. As a result, the His-tagged protein will finally be eluted with high recovery. Moreover, this product can be used for immunoprecipitation of His-tagged protein.


Generally the resin can be recycled at least four times. If maintained properly, the resin may be recycled 10 times with minor binding capability loss.

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR