Hematoksylin solution Gill III, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY

Synonymer: DcH-48, Natural Black I, Hydroxybrasilin

M-Clarity™ quality level = MQ500

GHS3128-4L
GHS3128-4LEA 3000 NOK
GHS3128-4L
Hematoksylin solution Gill III, Sigma-Aldrich®
Hematoksylin-fargeløsning

Gill formulation No. 3 can be used for cytology and/or histology as a progressive or regressive stain depending on length of staining time.


  • Used with hematoxylin and eosin staining
  • Contains 6 g/l certified hematoxylin


For in vitro diagnostic use.

Formel: C₁₆H₁₄O₆
MDL nummer: MFCD00078111
CAS-nummer: 517-28-2
IVDLogo

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR