ASPEC® 241 system
Leverandør: GILSON LCMS
26150006
GILI26150006EA 0 NOK
GILI26150006
ASPEC® 241 system
Ekstraktorer Automatiske ekstraksjonssystemer
This compact ASPEC® system combines positive pressure solid phase extraction (SPE) with small footprint and precise liquid handling for reliable automated SPE applications.
  • Positive pressure for reproducible extractions
  • Small footprint for minimised performance
  • Intuitive error handling for trusted results
  • Powerful software control for optimised productivity
  • Versatile set-up for trace analysis
With its minimised footprint, it is the ideal choice for basic SPE automation on 1, 3 or 6 ml cartridges for labs where bench space or throughput is limited. TheMore Product Information