Pipette holder, SINGLE™
Leverandør: GILSON
F161406
GILSF161406EA 225 NOK
GILSF161406
Pipette holder, SINGLE™
Pipettestativ
This convenient pipette holder has been designed to hold one single or multi channel pipette.
  • Made of PC: Resistant to aggressive chemicals and UV light
  • Adheres strongly to any smooth, dry surface or may be fastened with screws
  • Protects valuable instruments from shocks and contamination
  • Pipettes hang vertically to prevent liquids from running inside the shaft
  • Space saving: Adapted to limited work bench space