Pipette Stand for Multi-Channel Pipettes
Leverandør: GILSON
F161417
GILSF161417EA 470 NOK
GILSF161417
Pipette Stand for Multi-Channel Pipettes
Pipettestativ
The space-saving universal mulitchannel stand accommodates any brand of 8- or 12-channel pipette, protecting them from damage and contamination.
  • Compact stand optimizes bench space and fits more easily into fume hoods
  • Made from rugged materials that are resistant to many chemicals
The design and materials of the Universal Multichannel Pipette Stand allow the support edges to adapt to the lower part of virtually all pipettes. Because the stand holds a pipette vertically by its lower part, not the body as with most stands, the Universal Pipette Stand isMore Product Information