Ultra-Low Temperature Upright Freezer, VAULT100, Stirling Ultracold
Leverandør: Stirling Ultracold
VAULT100-EU
GLCLVAULT100-EUEA 0 NOK
GLCLVAULT100-EU
Ultra-Low Temperature Upright Freezer, VAULT100, Stirling Ultracold
Frysere Frysere, ultra-
The Stirling VAULT100 is the first ULT freezer with a −100 to −20 °C temperature range and the best steady-state setpoint adherence in the market.
  • ±3 °C* (9-position) uniformity and ±−0,2 °C* (single-position) stability at −80 °C
  • Improved M6D Stirling engine
  • Larger user interface with built-in ethernet port
  • Onboard predictive analytics
  • 6 year warranty
The Stirling VAULT100 features a new, more robust, free-piston Stirling engine design that delivers unsurpassed temperature uniformity and stability. It’s the first ULT freezer with a −100 to −20 °C