Cutlery, multi-purpose, Corning® Gosselin™
Leverandør: Corning
CFM-01
GOSSCFM-01EA 9675 NOK
GOSSCFM-01
Cutlery, multi-purpose, Corning® Gosselin™
Skjeer
Multi-purpose cutlery made from white PS. For sample collection and processing in food, pharmaceutical or cosmetic industries.
  • Food contact approved: Raw material is compliant with regulation EC N°1935/2004
  • Sterility assurance level based on ISO 11137: STERILE SAL 10⁻³
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904