Bottles, water sampling, octogonal, Corning® Gosselin™
Leverandør: Corning
P1000B-51 HP250B-53 P500B-51 P250B-51 HP500B-51 HP500B-50 HP250B-50 HP250B-51
331-0414EA 3225 NOK
331-0414 GOSSHP250B-53 GOSSP500B-51 GOSSP250B-51 331-0413 GOSSHP500B-50 GOSSHP250B-50 331-0412
Bottles, water sampling, octogonal, Corning® Gosselin™
Flasker Vannprøveflasker
The water samling bottles, made of PET, transparent, with screw cap are undosed or dosed with sodium thiosulphate, which inhibits the effects of chlorine, bromine and ozone present in the water when the sample is taken.
  • Molded graduations
  • Unbreakable
  • Tamper-evident screw cap
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat RECs, ISO 14001 - Borre