Portable Photometer
Leverandør: GRANT INSTRUMENTS
DEN-600
GRANDEN-600EA 12550 NOK
GRANDEN-600
Portable Photometer
Densitetsmålere
The DEN-600 is a compact, portable, rechargeable battery powered photometer for OD600, McF turbidity and Bradford protein assay methods.
  • An inexpensive alternative to a spectrophotometer, which is commonly used for these applications
  • Battery powered and compact, it can be comfortably located in a biosafety cabinet, anaerobic chamber or quickly relocated
  • The adjustable cell holder accommodates standard 10 mm path cuvettes, round bottom, conical vials or falcon tubes
  • USB connectivity and software allows data transfer, data processing and calculation