Test tubes with screw cap, sterile
Leverandør: Greiner Bio-One
164162_1000
GREI164162_1000EA 5275 NOK
GREI164162_1000
Test tubes with screw cap, sterile
Rør Reagensrør
Polystyrene (PS), clear.
  • Conical bottom, skirted
  • Good thermal resistance
  • Good chemical resistance
Universal tubes for a wide range of applications.