Cell culture tube, CELLSTAR®, CELLreactor™
Leverandør: Greiner Bio-One
227245_500
GREI227245_500EA 9425 NOK
GREI227245_500
Cell culture tube, CELLSTAR®, CELLreactor™
Rør Kulturrør
PP, graduated, sterile, with filter screw cap
  • Facilitates a high number of parallel experiments
  • Flexible working volume
  • Maximal sterility and excellent gas exchange
  • Conical tube design and in-tube harvest
The CELLSTAR® CELLreactor™ can be used as small bioreactor for the cultiviation of suspension cells. In addition to cell culture applications the CELLSTAR® CELLreactor™ can be applied for the expansion of aerobic bacteria, yeast or other microorganisms in shaken cultures* as well as storage of components and liquids requiring gas exchange.