Swabs in tubes
Leverandør: Greiner Bio-One
421161_1000 420161_1300 425010_1000
391-3795EA 5225 NOK
391-3795 GREI420161_1300 GREI425010_1000
Swabs in tubes
Svabere Svabere i rør
These swab tubes are suitable for taking non-human bacteriological, serological or cytological samples.
  • Sterilised by irradiation
  • Tubes are made of polystyrene
  • Swabs are made of cotton
Suitable for hygienic controls in food industry as well as in environmental sampling and veterinary research.
NB: *Produktet/produktene på denne siden er utenfor omfanget av EU -direktivet 2019/904
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001 (241002); ISO 50001 (241002)