VACUETTE® Blood Collection Tubes, without Additive
Leverandør: Greiner Bio-One
456085_1200
GREI456085_1200EA 2125 NOK
GREI456085_1200
VACUETTE® Blood Collection Tubes, without Additive
Diagnostiske systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Blood collection tubes made from practically unbreakable plastic
These tubes do not contain any additives and are used for CSF determinations or puncture specimens. Without threads, the tubes are especially suitable for automatic decapping by a quick pull movement.
They have made the drawing of blood a safer and more hygienic process. The use of PET plastic offers the advantage that the tubes are as clear as glass.