Blood collection tubes, with ACD, VACUETTE®
Leverandør: Greiner Bio-One
456094_1200
GREI456094_1200EA 0 NOK
GREI456094_1200
Blood collection tubes, with ACD, VACUETTE®
Diagnostiske systemer Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
These tubes are available with ACD solutions and are used for blood grouping determinations and also for cell preservation.