Biohazard Bags
Leverandør: Greiner Bio-One
649203_350
GREI649203_350EA 2725 NOK
GREI649203_350
Biohazard Bags
Poser Avfallsposer
PP disposal bag, with imprint 'Biohazard', for single use
  • Translucent bag with red printing
  • Suitable for steam autoclaves (max. 134 °C, 2 bar/30 min)
  • Temperature range: 0 to 140 °C