Cell culture plates, OneWell Plate™, CELLSTAR®
Leverandør: Greiner Bio-One
670180_32 670190_32
GREI670180_32EA 3725 NOK
GREI670180_32 GREI670190_32
Cell culture plates, OneWell Plate™, CELLSTAR®
Mikroplater Celledyrkningsplater
The CELLSTAR® OneWell Plate™ is designed for use in applications involving the cultivation of large quantities of cells. Made from PS.
  • Free from RNase, DNase, and human DNA
  • Non pyrogenic and non cytotoxic
The plate is suitable for use with a wide range of automated cell culture and liquid handling systems. The rectangular shape of the CELLSTAR® OneWell Plate™ allows for an efficient usage of incubator space. Notches on the left side of the plate and the lid ensure a secured lid position.
L×W×H: 127,8×85,5×14,4 mm
Sertifiseringer: Complies withMore Product Information