Adhesive seal, SILVERseal™
Leverandør: Greiner Bio-One
676090_1200
GREI676090_1200EA 19175 NOK
GREI676090_1200
Adhesive seal, SILVERseal™
Forseglinger Filmer og folier til mikroplateforsegling
SILVERseal™ is a heat resistant, cold-resistant and pierceable aluminium foil, which is suitable for both PCR applications and sample storage.
  • Temperature resistant from −80 to 110 °C
  • Coated with an acrylate adhesive, which remains flat when removed from its paper backing, and can be easily applied to all microplate types
  • A double perforation makes it possible to tear off the projecting adhesive sealer and thus improves the stacking of sealed microplates
VIEWseal™ is an adhesive sealing film coated with a silicone adhesive, which only sticks when theMore Product Information
Bestill nå
SPESIFIKASJONER
Environmentally Preferable Low Manufacturing Impact
Miljøsertifikat ISO 14001 (241002); ISO 50001 (241002)