Aspirating pipettes
Leverandør: Greiner Bio-One
710183_1000
GREI710183_1000EA 4600 NOK
GREI710183_1000
Aspirating pipettes
Pipetter Aspireringspipetter
PS pipettes, non plugged, non graduated, disposable, sterile.
  • Free of detectable DNase, RNase, human DNA
  • Non pyrogenic, non cytotoxic
  • Maximum accuracy, drop-free pipetting
  • Maximum clarity from high-grade polystyrene
  • Lot number and best-before date on each bag
Forpakning: Paper/plastic-packaging with peel-off function for individually wrapped pipettes with an additional break-through function.