Gel Load Pipette Tips
Leverandør: Greiner Bio-One
775390_10000
GREI775390_10000EA 7500 NOK
GREI775390_10000
Gel Load Pipette Tips
Pipettespisser
Gel-load pipette tips with a slim shape that allows even the thinnest gels to be loaded without damaging them. Due to the long and flexible tip, pipetting at the bottom of very narrow vessels is also possible without any problems.
  • Made of polypropylene
  • Universal fit for all common pipettes
  • 5 µl graduation for easy control of the pipetted volume
  • Tip with 0,57 mm outer diameter suitable for loading gel pockets
  • 83 mm total length to reach even the bottom of narrow vessels
The gel-load pipette tips are packed in a resealable bag, which means that the tipsMore Product Information