pH/mV/°C/rH meter, handheld, GMH 3531
Leverandør: Greisinger
602076
GRSG602076EA 0 NOK
GRSG602076
pH/mV/°C/rH meter, handheld, GMH 3531
Elektrokjemiske målere pH-meter
This meter is a simple handheld instrument for measuring pH, mV, temperature and relative humidity (0,0 to 70,0).
  • Battery operation makes it ideal for field measurements
  • Min./Max. value and HOLD memory
  • Impact-resistant ABS plastic housing, membrane keyboard, transparent panel; front side with IP 65 rating
  • Integrated pop-up clip for table top or suspended use
  • Serial interface (3,5 mm jack) can be connected to RS232 or USB interface of a PC via electrically isolated interface converter GRS 3100, GRS 3105 or USB 3100 N (available separately)
  • ORP mode allows
More Product Information
Bestill nå
SPESIFIKASJONER
Modell GMH 3531
Environmentally Preferable
pH - område 0,00 - 14,00
pH - oppløsning 0,01
pH - nøyaktighet ±0,01
pH - kalibrering 1-, 2- or 3- point
mV - område −1999...+2000; −1792...+2206 mVH (acc. to DIN 38404)
mV - oppløsning 1
mV - nøyaktighet ±0,1% FS
Temperaturområde −5,0...+150,0 °C
Temperaturoppløsning 0,1 °C
Temperaturnøyakighet ±0,2 °C (−5...+100 °C), otherwise ±0,4
temperaturkompensering Automatic (ATC) in the operating modes "pH" if temperature probe is used (0...105 °C); otherwise manual input
Display LCD (two lines, 2×4 digits, 12,4 + 7 mm height)
Strømforsyning 9 V battery, additional socket for external 10,5 - 12 V direct current power supply
Tilkoblinger BNC (pH, Redox, rH), 2 × 4 mm banana socket for Pt1000, 2-wire
Utganger Serial interface (3,5 mm jack); analog output
B×D×H 71×26×142 mm
Vekt 0,170 kg