Giemsa's stain modified, Sigma-Aldrich®

Leverandør: SIGMA ALDRICH MICROSCOPY
Danger

Synonymer: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

GS1L-1L
GS1L-1LEA 883 NOK
GS1L-1L
Giemsa's stain modified, Sigma-Aldrich®
Giemsa's stain

When blood films are stained using Giemsa Stain, the nucleus and cytoplasm of white blood cells take on characteristic blue or pink coloration. The use of purified eosin and thiazine dyes minimizes lot-to-lot variation.

Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00081642
CAS-nummer: 51811-82-6
ADR: 3 II
IVDLogo

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR