Ethanolamine benzoate hydrochloride ≥98%

Leverandør: Thermo Fisher Scientific
Warning

H25864.14
H25864.14EA 1725 NOK
H25864.14
Ethanolamine benzoate hydrochloride ≥98%
Ethanolamine benzoate hydrochloride

Purity: 98+% CAS No.: 545375-30-2 Molecular Formula: C9H11NO2.HCl Formula Weight: 201.65 Melting Point: 141-142° MDL No.: MFCD09953460

Formel: C₉H₁₁NO₂·HCl
MW: 201.65 g/mol
Smeltepunkt: 141…142 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD09953460
CAS-nummer: 545375-30-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR