2,2-Difluoropropionic acid ≥97%

Leverandør: Thermo Scientific
Danger

Synonymer: 2,2-Difluoropropanoic acid

H26030.03
H26030.03EA 4325 NOK
H26030.03
2,2-Difluoropropionic acid ≥97%
2,2-Difluoropropionic acid

Purity: 97% CAS No.: 373-96-6 Molecular Formula: C3F2H4O2 Formula Weight: 110.06 Flash Point: 66°(150°F) Melting Point: 36-44° Boiling Point: 142° UN No.: 3261 MDL No.: MFCD03093764

Formel: C₃H₄F₂O₂
MW: 110,06 g/mol
Kokepunkt: 142 °C
Smeltepunkt: 36…44 °C
Flash punkt: 66 °C (150 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD03093764
CAS-nummer: 373-96-6
FN: 3261
ADR: 8,II

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR