6-Chloro-m-toluidine ≥98%

Leverandør: Thermo Fisher Scientific
Danger

H26209.06 H26209.14
H26209.06EA 1475 NOK
H26209.06 H26209.14
6-Chloro-m-toluidine ≥98%
6-Chloro-m-toluidine

Purity: 98% CAS No.: 95-81-8 Molecular Formula: C7H8ClN Formula Weight: 141.60 Flash Point: 107°(225°F) Melting Point: 31-33° Boiling Point: 228-230° UN No.: 2239 MDL No.: MFCD00007674

Formel: C₇H₈ClN
Kokepunkt: 228…230 °C
Smeltepunkt: 31…33 °C
Flash punkt: 107 °C (225 °F)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00007674
CAS-nummer: 95-81-8
EINECS: 202-454-7
FN: 2239
ADR: 6.1,III

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR